missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Montage™ Plasmid Miniprep96 Kits
Generate high-quality plasmid DNA in 50% less time than traditional methods
£1415.00 - £6550.00
Specifications
Format | Microplate |
---|---|
For Use With (Application) | Plasmid Miniprep, BAC Prep |
Includes | All necessary reagents and disposables |
Product Type | Kit |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11854041
|
Merck
LSKP09604 |
4 x 96 Preps |
£1415.00
Pack of 4 |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11864041
|
Merck
LSKP09624 |
24 x 96 Preps |
£6550.00
Pack of 24 |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Typical yield from 1mL of bacterial culture is 4 to 6μg
- 96-well kit uses unique separation technology Simple, 25-minute protocol yields clean and reproducible DNA for sensitive downstream applications Collection of purified sample from the top of the plate and lack of centrifugation steps allow compatibility with automated systems
Specifications
Microplate | |
Plasmid Miniprep, BAC Prep | |
All necessary reagents and disposables | |
Kit |