missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera

Product Code. 11959064
Click to view available options
Grade:
Species: Group B, Antigen Factor 5
Species: Group B/D, Antigen Factors 12
Species: Group C, Antigen Factor 6 and 7
Species: Group E1, Antigen Factors 10
Species: Group E2, Antigen Factor 15
Species: Group E4, Antigen Factor 19
Species: Group G, Antigen Factors 13, 22 and 23
Species: Group G1, Antigen Factor 22
Species: Group G2, Antigen Factor 23
Species: Group K, Antigen Factor 18
Species: Group P, Antigen Factor 38
Unit Size:
3mL
Each
This item is not returnable. View return policy

Product Code. 11959064

Brand: Pro-Lab Diagnostics™ PL.6020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use in slide agglutination testing for identification of Salmonella species/antigens according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Somatic (O) Salmonella Antisera is a comprehensive range of monovalent Salmonella antisera delivered in economical 3.0mL bottles.

While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.

  • Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
  • Produced using reference strains prepared by World Health Organization guidelines
  • Certified reference center performance
  • Presented in color-coded dropper bottles
  • Long shelf life: up to 5 years from date of manufacture

Specifications

Certifications/Compliance W.H.O.
Content And Storage 2°C to 8°C, Do Not Freeze
Grade Species: Group E4, Antigen Factor 19
Product Type Antiserum
For Use With (Application) Traditional Slide Agglutination Testing
Packaging Type Dropper Bottle
Quantity 3.0 mL
Slide Type Agglutination
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.