missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera
Description
While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.
- Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
- Produced using reference strains prepared by World Health Organization guidelines
- Certified reference center performance
- Presented in color-coded dropper bottles
- Long shelf life: up to 5 years from date of manufacture
Specifications
Specifications
| Certifications/Compliance | W.H.O. |
| Content And Storage | 2°C to 8°C, Do Not Freeze |
| Grade | Antigen Factor z32 |
| Product Type | Antiserum |
| For Use With (Application) | Traditional Slide Agglutination Testing |
| Packaging Type | Dropper Bottle |
| Quantity | 3.0 mL |
| Slide Type | Agglutination |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?