missing translation for 'onlineSavingsMsg'
Learn More

Monosodium Glutamate Monohydrate, TRC

Product Code. 30471832
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471832 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471832 Supplier Toronto Research Chemicals Supplier No. TRCM5671505G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monosodium Glutamate Monohydrate
CAS 6106-04-3
Molecular Formula C5 H8 N O4 . H2 O . Na
Quantity 5 g
Synonym L-Glutamic acid, sodium salt, hydrate (1:1:1), Glutamic acid, monosodium salt, monohydrate, L- (8CI), L-Glutamic acid, monosodium salt, monohydrate (9CI), MSG monohydrate, Monosodium L-glutamate monohydrate, Monosodium glutamate monohydrate, Sodium glutamate monohydrate
InChI Formula InChI=1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1
SMILES O.[Na+].N[C@@H](CCC(=O)O)C(=O)[O-]
IUPAC Name sodium;(2S)-2-amino-5-hydroxy-5-oxopentanoate;hydrate
Molecular Weight (g/mol) 187.126
Formula Weight 187.046
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.