missing translation for 'onlineSavingsMsg'
Learn More

Monosialoganglioside GM3 Sodium Salt, TRC

Product Code. 30482420
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
Unit Size:
0.5mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30482420 1 mg 1mg
30482169 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30482420 Supplier Toronto Research Chemicals Supplier No. TRCM5671401MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monosialoganglioside GM3 Sodium Salt
Molecular Formula C59H107N2NaO21
Quantity 1 mg
Synonym Ganglioside GM3 Sodium Salt, GLac1 Sodium Salt, GM3 Sodium Salt, Ganglioside M3 Sodium Salt, Hematoside Sodium Salt, Sialosyllactosylceramide Sodium Salt
InChI Formula InChI=1S/C59H108N2O21.Na/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-46(69)61-40(41(66)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)38-77-56-51(73)50(72)53(45(37-64)79-56)80-57-52(74)55(49(71)44(36-63)78-57)82-59(58(75)76)34-42(67)47(60-39(3)65)54(81-59)48(70)43(68)35-62;/h30,32,40-45,47-57,62-64,66-68,70-74H,4-29,31,33-38H2,1-3H3,(H,60,65)(H,61,69)(H,75,76);/q;+1/p-1/b32-30+;/t40-,41+,42-,43+,44-,45-,47+,48+,49-,50+,51+,52+,53-,54+,55+,56+,57-,59?;/m0./s1
SMILES O[C@H]1CC(O[C@H]2[C@@H](O)[C@H](O[C@H]3[C@H](CO)O[C@@H](OC[C@]([C@@](O)([H])/C=C/CCCCCCCCCCCCC)([H])N([H])C(CCCCCCCCCCCCCCCCC)=O)[C@H](O)[C@H]3O)O[C@@H](CO)[C@@H]2O)(C(O[Na])=O)O[C@@H]([C@@]([H])([C@]([H])(O)CO)O)[C@@H]1NC(C)=O
Molecular Weight (g/mol) 1203.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.