missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl monopotassium malonate, 98%

Product Code. 10055723 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055723 500 g Glass bottle 500g
10180452 5 g Glass bottle 5g
10073240 25 g Glass bottle 25g
10177943 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10055723 Supplier Thermo Scientific Acros Supplier No. 269555000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38330-80-2 | C4H5O4·K | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38330-80-2
Molecular Formula C4H5O4·K
Molecular Weight (g/mol) 156.18
MDL Number MFCD00014021
InChI Key WWTULTKUWBKVGV-UHFFFAOYSA-M
Synonym potassium 3-methoxy-3-oxopropanoate, methyl potassium malonate, potassium methyl malonate, propanedioic acid, monomethyl ester, potassium salt, potassium monomethyl malonate, monomethyl malonate potassium salt, hydrogen methyl malonate potassium salt, monomethylpotassiummalonate, acmc-209vp4, ksc226a9d
PubChem CID 2724687
IUPAC Name potassium;3-methoxy-3-oxopropanoate
SMILES COC(=O)CC(=O)[O-].[K+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >200°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula COOKCH2COOCH3
Quantity 500 g
Fieser 15,207
Formula Weight 156.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl monopotassium malonate
Show More Show Less
compliance-icons
Product Identifier
  • Monomethyl monopotassium malonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.