missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Isophthalate, TRC

Product Code. 30471019
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471019 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471019 Supplier Toronto Research Chemicals Supplier No. TRCM52470010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monomethyl Isophthalate
CAS 1877-71-0
Molecular Formula C9H8O4
Quantity 10 g
Synonym 1-Methyl Ester-1,3-benzenedicarboxylic Acid, Monomethyl Ester-1,3-benzenedicarboxylic Acid, Methyl Ester Isophthalic Acid, Monomethyl Ester Isophthalic Acid, 3-(Methoxycarbonyl)benzoic Acid, 3-Carbomethoxybenzoic Acid, Methyl 3-Carboxybenzoate, Methyl Hydrogen Isophthalate, Methyl m-Carboxybenzoate, Methyl m-Phthalate
InChI Formula InChI=1S/C9H8O4/c1-13-9(12)7-4-2-3-6(5-7)8(10)11/h2-5H,1H3,(H,10,11)
SMILES COC(=O)c1cccc(c1)C(=O)O
Molecular Weight (g/mol) 180.16
Formula Weight 180.0423
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.