missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl auristatin E (MMAE) Antibody (08), Novus Biologicals™

Mouse Monoclonal Antibody

£164.00 - £343.00

Specifications

Antigen Monomethyl auristatin E (MMAE)
Clone 08
Dilution ELISA 1:1000-1:2000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30116704
View Documents
Novus Biologicals
NBP3-31974-100ug
100 μg
£343.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30115289
View Documents
Novus Biologicals
NBP3-31974-20ug
20 μg
£164.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Monomethyl auristatin E (MMAE) Monoclonal antibody specifically detects Monomethyl auristatin E (MMAE) in Nonspecies specific samples. It is validated for ELISA
Specifications

Specifications

Monomethyl auristatin E (MMAE)
ELISA 1:1000-1:2000
Monoclonal
Purified
RUO
0.2 um filtered solution in PBS
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Monomethyl auristatin E (MMAE) conjugated to BSA
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
08
ELISA
Unconjugated
Mouse
Non-species specific
474645-27-7, Monomethyl auristatin E (GMP), vedotin
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.