missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl Adipate, TRC

Product Code. 30478389
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478389 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478389 Supplier Toronto Research Chemicals Supplier No. TRCM56480025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monomethyl Adipate
CAS 627-91-8
Molecular Formula C7H12O4
Quantity 25 g
Synonym Hexanedioic Acid 1-Methyl Ester, Adipic Acid Methyl Ester, Adipic Acid Monomethyl Ester, Hexanedioic Acid Monomethyl Ester, 5-Carbomethoxypentanoic Acid, 6-Methoxy-6-oxohexanoic Acid, Methyl 5-Carboxypentanoate, Methyl Adipate, Methyl Hemiadipate, Methyl Hydrogen Adipate, Monomethyl 1,6-Hexanedioate, NSC 55113, NSC 9389
InChI Formula InChI=1S/C7H12O4/c1-11-7(10)5-3-2-4-6(8)9/h2-5H2,1H3,(H,8,9)
SMILES COC(=O)CCCCC(=O)O
Molecular Weight (g/mol) 160.17
Formula Weight 160.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.