missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl adipate, 97%

Product Code. 10439262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10439262 100 g Glass bottle 100g
10693702 5 g Glass bottle 5g
10011831 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10439262 Supplier Thermo Scientific Acros Supplier No. 102821000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-91-8 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-91-8
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00004418
InChI Key UOBSVARXACCLLH-UHFFFAOYSA-N
Synonym monomethyl adipate, adipic acid monomethyl ester, methyl hydrogen adipate, methyl adipate, methyl hemiadipate, hexanedioic acid, monomethyl ester, 5-carbomethoxypentanoic acid, monomethyl 1,6-hexanedioate, adipic acid, monomethyl ester, hexanedioic acid, 1-methyl ester
PubChem CID 12328
ChEBI CHEBI:70855
IUPAC Name 6-methoxy-6-oxohexanoic acid
SMILES COC(=O)CCCCC(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.0°C to 9.0°C
Color Colorless
Density 1.0800g/mL
Boiling Point 162.0°C (10.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HO2C(CH2)4CO2CH3
Refractive Index 1.4390 to 1.4420
Quantity 100 g
Beilstein 02, III, 1711
Specific Gravity 1.08
Formula Weight 160.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Monomethyl adipate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Monomethyl adipate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.