missing translation for 'onlineSavingsMsg'
Learn More

Monomelittoside, MedChemExpress

Product Code. 16401608
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401608 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401608 Supplier Medchem Express Supplier No. HYN0916

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Monomelittoside is a natural compound.

Monomelittoside is a natural compound.

Specifications

Chemical Name or Material Monomelittoside
CAS 20633-72-1
Purity Grade Notes Research
Molecular Formula C15H22O10
Quantity 5 mg
Synonym Danmelittoside
SMILES O[C@@]([C@@H]1O)(C=CO2)[C@](C(CO)=C1)([H])[C@@H]2O[C@]([C@@H]([C@@H](O)[C@@H]3O)O)([H])O[C@@H]3CO
Molecular Weight (g/mol) 362.33
Formula Weight 362.33
Grade Research
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.