missing translation for 'onlineSavingsMsg'
Learn More

Monoglyceride Lipase Antibody (OTI2B11), DyLight 594, Novus Biologicals™

Product Code. 18427212 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427212 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427212 Supplier Novus Biologicals Supplier No. NBP272759DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Monoglyceride Lipase Monoclonal antibody specifically detects Monoglyceride Lipase in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monoglyceride Lipase
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2B11
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias HU-K5EC 3.1.1.23, Lysophospholipase homolog, Lysophospholipase-like, MAGL, MGLHUK5, Monoacylglycerol lipase, monoglyceride lipase
Host Species Mouse
Immunogen Full length recombinant protein of human Monoglyceride Lipase (NP_009214) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 11343
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.