missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Monoglyceride Lipase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£162.00 - £386.00

Specifications

Antigen Monoglyceride Lipase
Dilution Western Blot : 1-2 μg/mL, Immunohistochemistry : 2-4 μg/mL
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066793
View Documents
Novus Biologicals™
NBP3-30968-25ug
25 μg
£162.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067229
View Documents
Novus Biologicals™
NBP3-30968-100ug
100 μg
£386.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Monoglyceride Lipase Polyclonal antibody specifically detects Monoglyceride Lipase in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications

Specifications

Monoglyceride Lipase
Immunohistochemistry, Western Blot
Unconjugated
Rabbit
Lipid and Metabolism
Phosphate buffered saline
11343
IgG
Antigen Affinity-purified
Western Blot : 1-2 μg/mL, Immunohistochemistry : 2-4 μg/mL
Polyclonal
Purified
RUO
Human
HU-K5EC 3.1.1.23, Lysophospholipase homolog, Lysophospholipase-like, MAGL, MGLHUK5, Monoacylglycerol lipase, monoglyceride lipase
Synthetic peptide derived from the human Monoglyceride Lipase protein.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.