missing translation for 'onlineSavingsMsg'
Learn More

Monoethyl Fumarate, Mikromol™

Product Code. 30049496
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049496 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049496 Supplier Mikromol Supplier No. MM1130.18

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Monoethyl Fumarate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Monoethyl Fumarate
CAS 5-4-2459
Analyte or Component Names Monoethyl Fumarate
Shipping Condition Room Temperature
Molecular Formula C6 H8 O4
Certifications/Compliance ISO 17025
Synonym (2E)-2-Butenedioic acid 1-ethyl ester, 2-Butenedioic acid (2E)-, monoethyl ester (9CI), 2-Butenedioic acid (E)-, monoethyl ester, Fumaric acid, ethyl ester (6CI,7CI), Fumaric acid, monoethyl ester (8CI), (E)-4-Ethoxy-4-oxo-2-butenoic acid, Ethyl hydrogen fumarate, Monoethyl fumarate, Monoethyl fumerate, Monoethyl trans-2-butenedioate
InChI Formula InChI=1S/C6H8O4/c1-2-10-6(9)4-3-5(7)8/h3-4H,2H2,1H3,(H,7,8)/b4-3+
SMILES CCOC(=O)\C=C\C(=O)O
IUPAC Name (E)-4-ethoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol) 144.13
Quantity 250 mg
Formula Weight 144.0423 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.