missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monodesethylamodiaquine Dihydrochloride, Mikromol™
Description
Monodesethylamodiaquine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | N-Desethylamodiaquine dihydrochloride |
| CAS | 79049-30-2 |
| Analyte or Component Names | N-Desethylamodiaquine dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 Cl N3 O . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Monodesethylamodiaquine Dihydrochloride |
| InChI Formula | InChI=1S/C18H18ClN3O.2ClH/c1-2-20-11-12-9-14(4-6-18(12)23)22-16-7-8-21-17-10-13(19)3-5-15(16)17;;/h3-10,20,23H,2,11H2,1H3,(H,21,22);2*1H |
| SMILES | Cl.Cl.CCNCc1cc(Nc2ccnc3cc(Cl)ccc23)ccc1O |
| IUPAC Name | 4-[(7-chloroquinolin-4-yl)amino]-2-(ethylaminomethyl)phenol;dihydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?