missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monocyte/Macrophage Antibody (KUL01) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28359
This item is not returnable.
View return policy
Description
Monocyte/Macrophage Monoclonal specifically detects Monocyte/Macrophage in Chicken samples. It is validated for Flow Cytometry.
Specifications
| Monocyte/Macrophage | |
| Monoclonal | |
| 0.5 mg/mL | |
| Flow Cytometry <= 1 ug/10^6 cells | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Chicken monocytes and macrophages | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry | |
| KUL01 | |
| Unconjugated | |
| macrophage, macrophages, monocyte, monocytes | |
| Chicken peripheral blood mononuclear cell leukocytes | |
| 0.5 mg | |
| Primary | |
| Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction