missing translation for 'onlineSavingsMsg'
Learn More

Monocrotaline, MedChemExpress

Product Code. 16449136
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16449136

Brand: Medchem Express HYN0750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Monocrotaline is an pyrrolizidine alkaloid extracted from the seeds of the Crotalaria spectabilis plant to induce pulmonary hypertension in rodents.

Monocrotaline is an pyrrolizidine alkaloid extracted from the seeds of the Crotalaria spectabilis plant to induce pulmonary hypertension in rodents.

Specifications

Chemical Name or Material Monocrotaline
CAS 315-22-0
Color White
Purity Grade Notes Research
Molecular Formula C16H23NO6
Quantity 100 mg
Synonym Crotaline
Solubility Information DMSO : 27.78 mg/mL (85.38 mM; Need ultrasonic) ∣H2O : 2 mg/mL (6.15 mM; Need ultrasonic)
SMILES O=C(O[C@]1([H])CCN2[C@]1([H])C(CO3)=CC2)[C@H](C)[C@@](C)(O)[C@@](C)(O)C3=O
Molecular Weight (g/mol) 325.36
Formula Weight 325.36
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.