Learn More
Heidolph™ MonoBlock Flask Insert
Product Code. 10497854 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
25 mL Flask
50 mL Flask
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
10497854 50 mL Flask Each
11781679 25 mL Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10497854 Supplier Heidolph™ Supplier No. 5058004200
Only null left
Add to basket
Add to basket
Accommodate specifically sized flasks at your StarFish magnetic stirrer workstation. Heidolph™ MonoBlock Flask Insert, a one-piece MonoBlock insert for flasks, is ideal for experiments using the same vessel.
  • Accommodates one specific flask size.
  • One-piece construction.
TRUSTED_SUSTAINABILITY
Specifications
Description Monoblock insert
For Use With (Equipment) Starfish Workstation, MR-Hei Magnetic Stirrers, 5 x 250 mL Flask MonoBlock
Holds 50 mL Flask