missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monoamine Oxidase B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528338
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Mouse (87%), Rat (86%), Dog (94%), Pig (90%), Bovine (90%), Guinea pig (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MAOB (Monoamine Oxidase B), also called MAO, BRAIN, AMINE OXIDASE (FLAVIN-CONTAINING) B, is a protein that in humans is encoded by the MAOB gene. MAOB is a member of the flavin monoamine oxidase family. And it is mapped on Xp11.3. MAOB catalyzes the oxidative deamination of biogenic and xenobiotic amines and plays an important role in the metabolism of neuroactive and vasoactive amines in the central nervous system and peripheral tissues. This protein preferentially degrades benzylamine and phenylethylamine. Like MAOA, it also degrades dopamine. MAO-B is involved in the breakdown of dopamine, a neurotransmitter implicated in reinforcing and motivating behaviors as well as movement. MAO-B inhibition is, therefore, associated with enhanced activity of dopamine, as well as with decreased production of hydrogen peroxide, a source of reactive oxygen species.
Specifications
| Monoamine Oxidase B | |
| Polyclonal | |
| Unconjugated | |
| MAOB | |
| 6330414K01Rik; adrenalin oxidase; amine oxidase [flavin-containing] B; HGNC:6834; MAO b; MAO, brain; MAO, platelet; Maob; MAO-B; MGC26382; monoamine oxidase B; monoamine oxidase type B; monoamine oxidase-B; RP1-201D17__B.1; tyramine oxidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109731, 25750, 4129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.51 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P19643, P27338, Q8BW75 | |
| MAOB | |
| Recombinant protein encompassing a sequence within the center region of human Monoamine Oxidase B. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction