missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoamine Oxidase B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35587-100ul
This item is not returnable.
View return policy
Description
Monoamine Oxidase B Polyclonal antibody specifically detects Monoamine Oxidase B in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| Monoamine Oxidase B | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| adrenalin oxidase, amine oxidase [flavin-containing] B, EC 1.4.3, EC 1.4.3.4, MAO, brain, MAO, platelet, MAO-B, MGC26382, monoamine oxidase B, Monoamine oxidase type B, tyramine oxidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 400-490 of human Monoamine Oxidase B (MAOB) (NP_000889.3).,, Sequence:, TYFPPGILTQYGRVLRQPVDRIYFAGTETATHWSGYMEGAVEAGERAAREILHAMGKIPEDEIWQSEPESVDVPAQPITTTFLERHLPSVP | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4129 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction