missing translation for 'onlineSavingsMsg'
Learn More

Monoamine Oxidase B Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NB100-2826-0.025mg

 View more versions of this product

Product Code. 18577383

  • £131.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Monoamine Oxidase B Polyclonal specifically detects Monoamine Oxidase B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monoamine Oxidase B
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
adrenalin oxidase, amine oxidase [flavin-containing] B, EC 1.4.3, EC 1.4.3.4, MAO, brain, MAO, platelet, MAO-B, MGC26382, monoamine oxidase B, Monoamine oxidase type B, tyramine oxidase
Goat
Affinity Purified
RUO
4129
Human
IgG
Western Blot
0.5 mg/mL
Western Blot 0.1 - 0.3 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:32000
NP_000889.3
MAOB
Peptide with sequence C-HKARKLARLTKEE corresponding to internal region according to NP_000889.3.
0.025 mg
Primary
This antibody is not expected to cross-react with MAOA.
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only