missing translation for 'onlineSavingsMsg'
Learn More

mono-Methyl isophthalate, 97%

Product Code. 10316464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316464 5 g Glass bottle 5g
10643995 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10316464 Supplier Thermo Scientific Acros Supplier No. 398230050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1877-71-0 | C9H8O4 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1877-71-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00029972
InChI Key WMZNGTSLFSJHMZ-UHFFFAOYSA-N
Synonym 3-methoxycarbonyl benzoic acid, mono-methyl isophthalate, methyl hydrogen isophthalate, isophthalic acid monomethyl ester, monomethyl isophthalate, isophthalic acid methyl ester, isophthalic acid, methyl ester, isophthalic acid, monomethyl ester, methyl m-phthalate, methyl 3-carboxybenzoate
PubChem CID 601880
IUPAC Name 3-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC(=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 194°C to 196°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (COOH)C6H4COOCH3
Quantity 5 g
Formula Weight 180.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material mono-Methyl isophthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.