missing translation for 'onlineSavingsMsg'
Learn More

mono-Methyl succinate, 95%

Product Code. 10103251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103251 25 g Glass bottle 25g
10431392 10 g Glass bottle 10g
10613522 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10103251 Supplier Thermo Scientific Acros Supplier No. 127730250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3878-55-5 | C5H8O4 | 132.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3878-55-5
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.11
MDL Number MFCD00002788
InChI Key JDRMYOQETPMYQX-UHFFFAOYSA-N
Synonym monomethyl succinate, succinic acid monomethyl ester, methyl hydrogen succinate, mono-methyl succinate, butanedioic acid, monomethyl ester, 3-carbomethoxypropanoic acid, succinic acid, monomethyl ester, unii-ya2v724s0a, mono-methyl hydrogen succinate, butanedioic acid monomethyl ester
PubChem CID 77487
ChEBI CHEBI:75146
IUPAC Name 4-methoxy-4-oxobutanoic acid
SMILES COC(=O)CCC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 55°C to 59°C
Color White
Boiling Point 151°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HO2CCH2CH2CO2CH3
Quantity 25 g
Beilstein 02, 608
Formula Weight 132.11
Percent Purity 95%
Physical Form Crystalline Chunks or Crystals
Chemical Name or Material mono-Methyl succinate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.