missing translation for 'onlineSavingsMsg'
Learn More

Mono-methyl phthalate, 98%

Product Code. 10025932 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10025932 100 g Glass bottle 100g
10128012 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10025932 Supplier Thermo Scientific Acros Supplier No. 374881000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4376-18-5 | C9H8O4 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4376-18-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
MDL Number MFCD00002466
InChI Key FNJSWIPFHMKRAT-UHFFFAOYSA-N
Synonym 2-methoxycarbonyl benzoic acid, methyl hydrogen phthalate, monomethyl phthalate, mono-methyl phthalate, phthalic acid monomethyl ester, o-methoxycarbonyl benzoic acid, monomethyl 1,2-benzenedicarboxylate, phthalic acid, monomethyl ester, monomethylphthalate, unii-2stt6d18jr
PubChem CID 20392
IUPAC Name 2-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 81.0°C to 86.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (COOH)C6H4COOCH3
Quantity 100 g
Solubility Information Solubility in water: insoluble (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material mono-Methyl phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.