missing translation for 'onlineSavingsMsg'
Learn More

mono-Ethyl succinate, 95%

Product Code. 10311580 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10311580 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10311580 Supplier Thermo Scientific Acros Supplier No. 436880050
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1070-34-4 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1070-34-4
CAS Min % 94.0
CAS Max % 100.0
Melting Point 8°C
Color Colorless
Density 1.1410g/mL
Elemental Analysis 46.4 to 52.3% C, 41.2 to 46.4% O
Boiling Point 248°C
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H10O4
Refractive Index 1.432
Quantity 5 g
Specific Gravity 1.141
Synonym monoethyl succinate, ethyl hydrogen succinate, mono-ethyl succinate, butanedioic acid, monoethyl ester, butanedioic acid, 1-ethyl ester, succinic acid monoethyl ester, monoethyl butanedioate, bernsteinsaureathylester, succinic acid monoethyl, succinic acid ethyl ester
InChI Key LOLKAJARZKDJTD-UHFFFAOYSA-N
SMILES CCOC(=O)CCC(=O)O
IUPAC Name 4-ethoxy-4-oxobutanoic acid
Molecular Weight (g/mol) 146.14
PubChem CID 70610
Formula Weight 146.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material mono-Ethyl succinate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • mono-Ethyl succinate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.