missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mono/Di-Methyl-Histone H3 (Lys9) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80219-1-RR-100UL
This item is not returnable.
View return policy
Description
The Mono/Di-Methyl-Histone H3 (Lys9) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Mono/Di-Methyl-Histone H3 (Lys9). This antibody recognizes human, mouse, rat antigen. The Mono/Di-Methyl-Histone H3 (Lys9) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| Mono/Di-Methyl-Histone H3 (Lys9) | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 8350 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1E12 | |
| Unconjugated | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 15-18 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction