missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus I1L Antibody [DyLight 405], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43764V
This item is not returnable.
View return policy
Description
Monkeypox Virus I1L Polyclonal antibody specifically detects Monkeypox Virus I1L in Virus samples. It is validated for Western Blot
Specifications
| Monkeypox Virus I1L | |
| Polyclonal | |
| Western Blot | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50 mM Sodium Borate | |
| Produced in rabbits immunized with purified, recombinant Monkeypox Virus I1L (Met1-Glu312) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction