missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus B6R (strain Zaire-96-I-16) Recombinant Rabbit Monoclonal Antibody (HL3107)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556797
This item is not returnable.
View return policy
Description
Monkeypox Virus B6R (strain Zaire-96-I-16) Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. MPXV A29L, M1R and B6R, the orthologous to A27L, L1R and B5R VACV antigens respectively, are thought to be able to elicit a protective immune response against lethal MPXV challenge.
Specifications
| Monkeypox Virus B6R (strain Zaire-96-I-16) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL3107 | |
| Unconjugated | |
| Monkeypox B6R | |
| Recombinant fragment of Mpox virus B6R.(#MPXV_USA_2022_MA001). | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction