missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISA (Matched Antibody Pair)
Specifications
Specifications
| Antigen | Monkeypox Virus A29L |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 31 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:5000, ELISA, ELISA Capture (Matched Antibody Pair) 1:250-1:2000 |
| Formulation | 0.2 um filtered solution in PBS |
| Host Species | Monkey |
| Immunogen | Recombinant Monkeypox Virus A29L Protein |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?