Learn More
Invitrogen™ ICAM-1 (Soluble) Monkey ELISA Kit
For the quantitative detection of monkey sICAM-1.
Brand: Invitrogen™ BMS648
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sICAM-1
HRP-Conjugate anti-monkey sICAM-1 monoclonal antibody
Monkey sICAM-1 Standard, 100 U/ml
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
Description
The Monkey Soluble Intercellular Adhesion Molecule-1 (ICAM-1) ELISA quantitates Mk ICAM-1 in monkey serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Mk ICAM-1. Principle of the method The Monkey ICAM-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.Specifications
Q5NKV6 | |
1.1 U/mL | |
ELISA Kit | |
Monkey | |
Colorimetric Microplate Reader | |
ICAM1 | |
5% | |
HRP | |
RUO | |
2°C to 8°C | |
Baboon, Rhesus Monkey | |
1 hr. 10 min. |
6.3-100 U/mL | |
HRP | |
Serum, Supernatant | |
ELISA | |
101021731, 712280 | |
10% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers | |
96 Tests | |
Serum, 10 μL; Supernatant, 10 μL | |
Intercellular adhesion molecule 1, CD54 antigen | |
1 hr. 20 min. |
For Research Use Only