missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-selectin (Soluble) Monkey ELISA Kit

Product Code. p-7092141
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
96 tests
Pack of 10
This item is not returnable. View return policy

Product Code. 15532097

Brand: Invitrogen™ BMS649

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sE-selectin
  • HRP-Conjugate anti-monkey sE-selectin monoclonal antibody
  • Monkey sE-selectin Standard lyophilized, 120 U/ml upon reconstitution
  • Sample Diluent
  • Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
  • Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Blue-Dye
  • Green-Dye
  • Adhesive Films

For the quantitative detection of monkey sE-selectin.

The Monkey Endothelial Leukocyte Adhesion Molecule (Mk e-Selectin/ELAM-1) ELISA quantitates Mk ELAM-1 in baboon serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Mk ELAM-1. Principle of the method The Monkey ELAM-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

E-selectin (CD62E, ELAM-1, and LECAM-1) is a 115 kD type I membrane protein and a member of the selectin family. E-selectin is highly expressed on activated endothelial cells (IL-2, TNF- alpha, other cytokines can increase expression) and can also be expressed on endothelial cells in the skin, bone marrow and placenta. E-selectin is involved in tethering and leucocyte rolling on activated endothelium at inflammatory sites and may also play a role in tumor metastasis and angiogenesis. E-selectin binds to both Siayl Lewis X (CD15s) and PSGL-1 (CD162). The HCD62E antibody has been shown to recognize human E-selectin and to be useful for flow cytometry. In vitro experiments have shown that IL-1, TNFalpha and bacterial wall components like lipopolysaccharides induce the transcription of E-selectin in a NFkB dependent signaling cascade. E-selectin has been associated with blood vessel endothelium in diverse inflammatory situations.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 1.9-60 U/mL
Assay Sensitivity 0.4 U/mL
Conjugate HRP
Product Type ELISA Kit
Sample Type Serum, Supernatant
Target Species Monkey
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 101009590
Gene Symbol SELE
Interassay CV 10%
Intraassay CV 5%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Serum, 20 μL; Supernatant, 20 μL
Storage Requirements 2°C to 8°C
Synonym ELAM1, endothelial adhesion molecule 1, CD62E
Target Baboon
Test Time 1 hr. 20 min.
Total Assay Time 2 hr. 10 min.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.