missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Monkey PD-L1/B7-H1 DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
Pack of 15
Specifications
Specifications
| Assay Range | 0.31 to 20 ng/mL |
| Product Type | Solid Phase Sandwich ELISA |
| Target Species | Primate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 29126 |
| Quantity | 1 Kit (for 15 Plates) |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 |
| Target | PD-L1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction