missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkey Osteoactivin/GPNMB ELISA Kit

Product Code. 17194943
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17194943

Brand: Invitrogen™ EP24RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Monkey Osteoactivin/GPNMB quantitates monkey Osteoactivin/GPNMB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey Osteoactivin/GPNMB.

The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; Gpnmb; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; HGFIN; ipd; iris pigment dispersion; NMB; Osteoactivin; transmembrane glycoprotein HGFIN; Transmembrane glycoprotein NMB; UNQ1725/PRO9925
Assay Range 0.12 to 25 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target GPNMB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.