missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Monkey FGF-21 ELISA Kit (Colorimetric)

Product Code. 18384043
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384043 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384043 Supplier Novus Biologicals™ Supplier No. NBP318048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25-2000 pg/mL
Assay Sensitivity 18.75 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Biological Fluids, Plasma, Serum
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 26291
Gene Symbol FGF21
Quantity 1 Kit
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym FGF-21, fibroblast growth factor 21
Target Monkey
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.