missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Moniliformin sodium salt

Product Code. 16470939
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470939 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16470939 Supplier MedChemExpress Supplier No. HY101905
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Moniliformin sodium salt is a potent mycotoxin isolate from Fusarium moniliforme.

Specifications

Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Molecular Weight (g/mol) 120.04 g/mol
Shipping Condition Room temperature
For Use With (Application) COVID-19-immunoregulation
CAS 71376-34-6
Product Type Natural Products
Purity 99.35%
Solubility H2O : 100 mg/mL (833.06 mM∣Need ultrasonic) ∣DMSO : ≥ 45 mg/mL (374.88 mM)
Target Fungal∣Antibiotic
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.