missing translation for 'onlineSavingsMsg'
Learn More

Monic Acid A, TRC

Product Code. 30132932
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132932 50 mg 50mg
30011471 10 mg 10mg
30003289 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132932 Supplier Toronto Research Chemicals Supplier No. TRCM52050050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66262-68-8
Molecular Formula C17H28O7
Molecular Weight (g/mol) 344.4
Synonym L-talo-Non-2-enonic acid, 5,9-anhydro-2,3,4,8-tetradeoxy-8-[[(2S,3S)-3-[(1S,2S)-2-hydroxy-1-methylpropyl]oxiranyl]methyl]-3-methyl-, (2E)- (9CI, ACI), (2E)-5,9-Anhydro-2,3,4,8-tetradeoxy-8-[[(2S,3S)-3-[(1S,2S)-2-hydroxy-1-methylpropyl]oxiranyl]methyl]-3-methyl-L-talo-non-2-enonic acid (ACI), L-talo-Non-2-enonic acid, 5,9-anhydro-2,3,4,8-tetradeoxy-8-[[3-(2-hydroxy-1-methylpropyl)oxiranyl]methyl]-3-methyl-, [2E,8[2S,3S(1S,2S)]]- (ZCI), Monic acid, Monic acid A
IUPAC Name (E)-4-[(2S,3R,4R,5S)-3,4-dihydroxy-5-[[(2S,3S)-3-[(2S,3S)-3-hydroxybutan-2-yl]oxiran-2-yl]methyl]oxan-2-yl]-3-methylbut-2-enoic acid
SMILES C[C@H](O)[C@H](C)[C@@H]1O[C@H]1C[C@H]2CO[C@@H](C\C(=C\C(=O)O)\C)[C@H](O)[C@@H]2O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H28O7/c1-8(5-14(19)20)4-12-16(22)15(21)11(7-23-12)6-13-17(24-13)9(2)10(3)18/h5,9-13,15-18,21-22H,4,6-7H2,1-3H3,(H,19,20)/b8-5+/t9-,10-,11-,12-,13-,15+,16-,17-/m0/s1
Formula Weight 344.1835
Chemical Name or Material Monic Acid A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.