missing translation for 'onlineSavingsMsg'
Learn More

Monastrol, MedChemExpress

Product Code. 16469796
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469796 50 mg 50mg
16465977 5 mg 5mg
16410729 1 mg 1mg
16403258 10 mg 10mg
16402696 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16469796 Supplier Medchem Express Supplier No. HY101071A
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Monastrol is a potent and cell-permeable inhibitor of the mitotic kinesin Eg5 with an IC50 value of 14 μM.

Monastrol is a potent and cell-permeable inhibitor of the mitotic kinesin Eg5 with an IC50 value of 14 μM.

Specifications

Chemical Name or Material Monastrol
CAS 329689-23-8
Color White
Purity Grade Notes Research
Molecular Formula C14H16N2O3S
Quantity 50 mg
Synonym (±)-Monastrol
Solubility Information DMSO : ≥ 33 mg/mL (112.88 mM)
SMILES O=C(C1=C(C)NC(NC1C2=CC=CC(O)=C2)=S)OCC
Molecular Weight (g/mol) 292.35
Formula Weight 292.35
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.