missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MON1A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563511

Product Code. 15791282

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VALVSFLEAD KNAIRSIHAD GYKVVFVRRS PLVLVAVART RQSAQELAQE LLYIYYQILS LLTGAQLSHI FQQKQN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

MON1A is a cytosolic protein that peripherally associates with the ER. MON1A plays an important role in membrane trafficking through the secretory apparatus. MON1A affects different steps in the secretory pathway including endoplasmic reticulum-to-Golgi traffic. MON1A encodes several isoforms with MW about 62, 55, 45 and 34 kDa.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MON1A
Polyclonal
Unconjugated
MON1A
2810468K17Rik; FLJ97088; MGC13272; MON1 homolog A; MON1 homolog A (yeast); MON1 homolog A, secretory trafficking associated; MON1 secretory trafficking family member A; Mon1a; RGD1308823; SAND1; Vacuolar fusion protein MON1 homolog A
Rabbit
Antigen affinity chromatography
RUO
84315
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q86VX9
MON1A
Recombinant protein corresponding to Human MON1A. Recombinant protein control fragment (Product #RP-96427).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.