missing translation for 'onlineSavingsMsg'
Learn More

Momelotinib sulfate, MedChemExpress

Product Code. 16464318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464318 10 mg 10mg
16457037 5 mg 5mg
16425236 100 mg 100mg
16420647 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16464318 Supplier Medchem Express Supplier No. HY10962
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Momelotinib sulfate (CYT387 sulfate salt) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, 10-fold selectivity versus JAK3 (IC50=155 nM).

Momelotinib sulfate (CYT387 sulfate salt) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, 10-fold selectivity versus JAK3 (IC50=155 nM).

Specifications

Chemical Name or Material Momelotinib sulfate
CAS 1056636-06-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H26N6O10S2
Quantity 10 mg
Synonym CYT387 sulfate salt
Solubility Information DMSO : 220 mg/mL (360.29 mM; Need ultrasonic) ∣H2O : 100 mg/mL (163.77 mM; Need ultrasonic)
SMILES O=C(NCC#N)C(C=C1)=CC=C1C2=NC(NC(C=C3)=CC=C3N4CCOCC4)=NC=C2.O=S(O)(O)=O.O=S(O)(O)=O
Molecular Weight (g/mol) 610.62
Formula Weight 610.62
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.