missing translation for 'onlineSavingsMsg'
Learn More

Momelotinib sulfate, MedChemExpress

Product Code. 16425236
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425236

Brand: Medchem Express HY10962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Momelotinib sulfate (CYT387 sulfate salt) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, 10-fold selectivity versus JAK3 (IC50=155 nM).

Momelotinib sulfate (CYT387 sulfate salt) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50 of 11 nM/18 nM, 10-fold selectivity versus JAK3 (IC50=155 nM).

Specifications

Chemical Name or Material Momelotinib sulfate
CAS 1056636-06-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H26N6O10S2
Quantity 100 mg
Synonym CYT387 sulfate salt
Solubility Information DMSO : 220 mg/mL (360.29 mM; Need ultrasonic) ∣H2O : 100 mg/mL (163.77 mM; Need ultrasonic)
SMILES O=C(NCC#N)C(C=C1)=CC=C1C2=NC(NC(C=C3)=CC=C3N4CCOCC4)=NC=C2.O=S(O)(O)=O.O=S(O)(O)=O
Molecular Weight (g/mol) 610.62
Formula Weight 610.62
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.