missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Momelotinib, MedChemExpress
Description
Momelotinib (CYT387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50a of 11 nM and 18 nM,respectively. CYT387 shows much less activity against JAK3.
Specifications
Specifications
| Chemical Name or Material | Momelotinib |
| CAS | 1056634-68-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H22N6O2 |
| Quantity | 200 mg |
| Synonym | CYT387 |
| Solubility Information | DMSO : 20.83 mg/mL (50.26 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(C1=CC=C(C2=NC(NC3=CC=C(N4CCOCC4)C=C3)=NC=C2)C=C1)NCC#N |
| Molecular Weight (g/mol) | 414.46 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?