missing translation for 'onlineSavingsMsg'
Learn More

Momelotinib, MedChemExpress

Product Code. 16437037
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437037

Brand: Medchem Express HY10961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Momelotinib (CYT387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50a of 11 nM and 18 nM,respectively. CYT387 shows much less activity against JAK3.

Momelotinib (CYT387) is an ATP-competitive inhibitor of JAK1/JAK2 with IC50a of 11 nM and 18 nM,respectively. CYT387 shows much less activity against JAK3.

Specifications

Chemical Name or Material Momelotinib
CAS 1056634-68-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H22N6O2
Quantity 5 mg
Synonym CYT387
Solubility Information DMSO : 20.83 mg/mL (50.26 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C1=CC=C(C2=NC(NC3=CC=C(N4CCOCC4)C=C3)=NC=C2)C=C1)NCC#N
Molecular Weight (g/mol) 414.46
Formula Weight 414.46
Percent Purity 96.95%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.