missing translation for 'onlineSavingsMsg'
Learn More

Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals

Catalog Number 11342598
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
7782-91-4
H2MoO4
161.96
MFCD00036287
VLAPMBHFAWRUQP-UHFFFAOYSA-L
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
82208
CHEBI:25371
dihydroxy(dioxo)molybdenum
O[Mo](O)(=O)=O
This item is not returnable. View return policy
7782-91-4
H2MoO4
161.96
MFCD00036287
VLAPMBHFAWRUQP-UHFFFAOYSA-L
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
82208
CHEBI:25371
dihydroxy(dioxo)molybdenum
O[Mo](O)(=O)=O

Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in dilute ammonium hydroxide,alkali and ammonia. Slightly soluble in water.
Grade ACS
Concentration or Composition (by Analyte or Components) MoO3 85% min.
Odor Odorless
Density 3.1g/mL
Chemical Name or Material Molybdic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'