missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide bis(2,4-pentanedionate), 99%

Product Code. p-7059180
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11309638

Brand: Thermo Scientific Alfa Aesar 044140.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17524-05-9 | C10H14MoO6 | 326.17 g/mol

Forms a functional model compound for the study of oxotransferase molybdenum enzymes. Mild catalyst for the deprotection of acetals in high yield. Catalyzes high yield oxidation of secondary alcohols to ketones with sodium percarbonate and phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17524-05-9
Molecular Formula C10H14MoO6
Molecular Weight (g/mol) 326.17
MDL Number MFCD00011506
InChI Key SKMUJBBRXZPAJY-VGKOASNMSA-L
Synonym Bis(acetylacetonato)dioxomolybdenum(VI); Molybdenyl acetylacetonate
IUPAC Name dioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
SMILES O=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼185°C (decomposition)
Color Yellow
Physical Form Powder
Quantity 2 g
Formula Weight 326.16
Percent Purity 99%
Sensitivity Air Sensitive
Chemical Name or Material Molybdenum (VI) oxide bis (2,4-pentanedionate)
compliance-icons
Product Identifier
  • Molybdenum(VI) oxide bis(2,4-pentanedionate)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Dioxobis(pentane-2,4-dionato-O,O)molybdenum

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.