missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W)

Product Code. 11362018
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362018 500 g 500g
11352018 25 g 25g
11342018 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362018 Supplier Thermo Scientific Alfa Aesar Supplier No. 012930.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

It is used as a selective oxidation catalyst, and as a molybdenum oxidase enzyme. It is a source of molybdenum compounds and molybdenum metal which serves as an additive to steel and corrosion-resistant alloys. It is used for lubricants and lubricant additives. It has applications in process regulators, pigments, corrosion inhibitors, electrochemical devices, displays, and smoke suppressants. It finds use in analytical chemistry, ceramic glazes, enamels, energy storage, and in the manufacture of acrylonitrile through catalytic oxidation of ammonia and propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C (sublimation)
Melting Point 795°C
Color White to Gray
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9995% (metals basis excluding W)
Solubility Information Slightly soluble in water. Soluble in concentrated mineral acids,alkali hydroxide solutions.
Formula Weight 143.94
Odor Odorless
Appearance White to gray powder
Density 4.692
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.