missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, 99.95% (metals basis)

Product Code. 11331898
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331898 2 kg 2kg
11311898 100 g 100g
11321898 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11331898 Supplier Thermo Scientific Alfa Aesar Supplier No. 011837.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

Molybdenum(VI) oxide is a very promising and stable molybdenum oxide for the electrochromic applications and used as a catalyst in oil industries. It is also used to make metal molybdenum. It has a wide band gap n-type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties. It has been widely investigated as a smart material for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials. Molybdenum(VI) oxide (MoO3) is employed as an adhesive between enamels and metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C (sublimation)
Melting Point 795°C
Color White
Physical Form Sublimed Powder
Quantity 2 kg
Assay Percent Range 99.95% (metals basis)
Solubility Information Barely soluble in water; soluble in base.
Formula Weight 143.94
Odor Odorless
Appearance White to gray powder
Density 4.692
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.