Learn More
Molybdenum(VI) oxide, 99.5%, Thermo Scientific Chemicals
Description
Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Molybdenum(VI) oxide is a very promising and stable oxide for electrochromic applications and used as a catalyst in the oil industry. It is also used to make metal molybdenum. It has a wide band gap type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties, has been widely investigated as smart materials for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials.
Solubility
Slightly soluble in water. Soluble in concentrated mineral acids and alkali hydroxide solutions.
Notes
It does not react with acids other than hydrofluoric and concentrated sulfuric acids.
Specifications
Boiling Point | 1155°C (sublimation) |
Melting Point | 795°C |
Quantity | 250 g |
UN Number | UN3288 |
Merck Index | 14,6239 |
Solubility Information | Slightly soluble in water. Soluble in concentrated mineral acids and alkali hydroxide solutions. |
Formula Weight | 143.94 |
Percent Purity | 99.5% |
Odor | Odorless |
Density | 4.692 |
Show More |
RUO – Research Use Only