missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) fluoride

Product Code. 15421097
Click to view available options
Quantity:
50 g
Packaging:
CGA 330 valve cylinder
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 15421097

Brand: Thermo Scientific Alfa Aesar 022713.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7783-77-9 | F6Mo | 209.94 g/mol

Molybdenum(VI) fluoride plays an important role in the semiconductor industry and the nuclear industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packed in one-way cylinder w/CGA 330 valve
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 37°C
Melting Point 17°C
CAS 7783-77-9
Color Colorless
Physical Form Liquid
Quantity 50 g
Molecular Formula F6Mo
Linear Formula MoF6
MDL Number MFCD00014220
UN Number UN3289
Merck Index 14,6237
Synonym molybdenum hexafluoride, molybdenum fluoride mof6 , oc-6-11, molybdenum fluoride mof6, unii-zlt5j123w3, mof6, molybdenum vi fluoride, molybdenum 6+ fluoride, acmc-1bii8
Solubility Information Miscible with saturated hydrocarbons,chlorinated and fluorinated hydrocarbons.
InChI Key RLCOZMCCEKDUPY-UHFFFAOYSA-H
SMILES F[Mo](F)(F)(F)(F)F
IUPAC Name hexafluoromolybdenum
Molecular Weight (g/mol) 209.94
PubChem CID 82219
ChEBI CHEBI:30708
Formula Weight 209.93
Packaging CGA 330 valve cylinder
Sensitivity Moisture sensitive
Density 2.3 g/mL
Chemical Name or Material Molybdenum(VI) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.