missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) trichloride-isopropoxide, 25% (w/v) in isopropanol, Thermo Scientific™

Product Code. 11410810
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410810 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11410810 Supplier Thermo Scientific Alfa Aesar Supplier No. 044474.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53780-81-7
Molecular Formula C6H16Cl3MoO2
Molecular Weight (g/mol) 322.492
MDL Number MFCD02682829
InChI Key JVWWBSIISOXHDE-UHFFFAOYSA-K
Synonym molybdenum v trichloride-isopropoxide, in isopropanol
PubChem CID 121233634
IUPAC Name propan-2-ol;trichloromolybdenum
SMILES CC(C)O.CC(C)O.Cl[Mo](Cl)Cl

Specifications

Quantity 50 mL
UN Number UN2924
Sensitivity Moisture Sensitive
Formula Weight 320.48
Concentration or Composition (by Analyte or Components) 25% (w/v) in isopropanol
Physical Form Liquid
Chemical Name or Material Molybdenum (V) trichloride-isopropoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.