missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) isopropoxide, 99+% (metals basis)

Product Code. 11324228
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324228 10 g 10g
11334228 0.5 g 0.5g
11314228 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324228 Supplier Thermo Scientific Alfa Aesar Supplier No. 041484.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Precursor used in sol-gel process | CAS: 209733-38-0 | C15H35MoO5 | 391.39 g/mol

Molybdenum(V) isopropoxide, it is used as a Precursor used in sol-gel process. It is used in nanomaterials and thin films. It is used as a solution based product for printed electronics. It acts a catalyst for selective isobutane oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209733-38-0
Molecular Formula C15H35MoO5
Molecular Weight (g/mol) 391.39
MDL Number MFCD00210636
InChI Key HLFWWCCBFJUYJL-UHFFFAOYSA-N
Synonym molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
PubChem CID 18475294
IUPAC Name molybdenum;propan-2-olate
SMILES CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Black to Yellow
Linear Formula Mo[OCH(CH3)2]5
Quantity 10 g
UN Number UN3181
Sensitivity Moisture Sensitive
Formula Weight 391.04
Percent Purity ≥99%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Molybdenum(V) isopropoxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.